missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Whole Serum, Bovine, Purified, 2mL
Whole Bovine Serum with buffer salts
Brand: MP Biomedicals™ 0855980
Additional Details : Weight : 0.00200kg
Specifications
2° to 8°C | |
Lyophilized | |
2 mL | |
Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques, used as a blocking agent in fluorescence and enzyme immunoassays |
Bovine | |
Adult Bovine Serum | |
0.2 μm filtration | |
61.9g/mL |