missing translation for 'onlineSavingsMsg'
Learn More

Welch™ MPC 090 E Vacuum Pump

Use this vacuum pump for sample preparation in life sciences, environmental monitoring, chemistry and analytical processes. Welch™ MPC 090 E Vacuum Pumps feature reliable diaphragm technology, ensuring quiet operation and long service life.

Brand:  Welch™ 412021

Product Code. 12625635

  • 1176.60 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • Standard high accuracy regulator prevents damage to the filter membrane, andchieves a powerful final vacuum level for fast results
  • Built-in catch pot prevents ingestion of liquid into the pump
  • Controllable filtration power
  • Compact, oil free, portable
  • Vacuum display and adjustment for soft start
  • Chemical duty pump
  • DN 6 hose nozzle Single funnel vacuum filtration, vacuum desiccators, nano-filtration, HPLC-filtration.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

15 L/min.
198 mm
2.7 kg
10°C
50/60 Hz
144 mm
100 mbar
260 V
MPC 090 E
40°C
15 L/min.
244 mm
Vacuum Pump
260 V
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Welch™ MPC 090 E Vacuum Pump > Depth: 198mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.