missing translation for 'onlineSavingsMsg'
Learn More

Welch™ MPC 104 Tp Vacuum Pump

For non-volatile solvents and acids

Brand:  Welch™ 412443-02

1749.86 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 11856281

  • 2235.00 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Tp flow rate 15L/min for non-volatile solvents and acids 230V 50/60Hz 235mm(w) x 140mm(d) x 340mm(h) vacuum >2mbar Ilmvac
Specifications

Specifications

230 V 50/60 Hz
MPC 104 T
15 L/min.
340 mm
Vacuum Pump
230 V
140 mm
7.1 kg
50/60 Hz
235 mm
<2 mbar
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.