Learn More
Welch™ MPC 301 Zp, ATEX Cat.2 Diaphragm Pump
Designed for use as a backing pump for rotary evaporators, vacuum ovens or gel dryer. Welch Ilmvac™ MPC 301 Zp Diaphragm Pumps are ideal for vacuum filtration, vacuum concentrating, distillation and desiccation.
Brand:  Welch™ 4000481-04
Product Code. 10091832
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • ATEX Cat.2 explosion proof diaphragm pump
  • Manual Vacuum regulator with dial gauge and liquid trap
  • Powerful corrosion resistant diaphragm vacuum pump build-in
  • Maintenance-free drive system and proven long diaphragm life
  • Requires minimal benchtop space due to compact design
  • Analytically pure, oil free vacuum
  • Light weight, rugged design
  • All wetted parts are made of Ryton, Viton or Kalrez
  • Designed for continuous running
  • Use in the lab or in the field (with optional adapter)
  • CE Marked
Specifications
Specifications
400 V 50 Hz
MPC 301 Zp, ATEX Cat.2
40°C
38 L/min.
260 mm
Diaphragm Pump
400 V
300 mm
22.9 kg
10°C
50 Hz
240 mm
<8 mbar