Learn More
MPPE1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15313-1-AP-20UL
 View more versions of this product
Product Code. 16834444
  • 145.00 € / 20µL
Explore more special offers
Description
Description
Metallophosphoesterase required for transport of GPI- anchor proteins from the endoplasmic reticulum to the Golgi. Acts in lipid remodeling steps of GPI-anchor maturation by mediating the removal of a side-chain ethanolamine-phosphate (EtNP) from the second Man (Man2) of the GPI intermediate, an essential step for efficient transport of GPI-anchor proteins.
Specifications
Specifications
MPPE1
Polyclonal
Unconjugated
MPPE1
metallophosphoesterase 1, MPPE1, PGAP5
Rabbit
Antigen Affinity Chromatography
RUO
225651, 361344, 65258
-20°C
Liquid
Western Blot
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
B1WC86, Q53F39, Q80XL7
Mppe1
MPPE1 Fusion Protein Ag7547
20 μL
Primary
Mouse, Human, Rat