missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MPS1 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518092
This item is not returnable.
View return policy
Description
This antibody is predicted to react with mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
Ribosomes, the organelles that catalyze protein synthesis, consist of a small 40S subunit and a large 60S subunit. Together these subunits are composed of 4 RNA species and approximately 80 structurally distinct proteins. This gene encodes a ribosomal protein that is a component of the 40S subunit. The protein belongs to the S27E family of ribosomal proteins. It contains a C4-type zinc finger domain that can bind to zinc. The encoded protein has been shown to be able to bind to nucleic acid. It is located in the cytoplasm as a ribosomal component, but it has also been detected in the nucleus. Studies in rat indicate that ribosomal protein S27 is located near ribosomal protein S18 in the 40S subunit and is covalently linked to translation initiation factor eIF3. As is typical for genes encoding ribosomal proteins, there are multiple processed pseudogenes of this gene dispersed through the genome.
Specifications
| MPS1 | |
| Polyclonal | |
| Unconjugated | |
| RPS27 | |
| 3200001M24Rik; 40S ribosomal protein S27; metallopanstimulin 1; Metallopan-stimulin 1; MPS1; MPS-1; RGD:621045}; ribosomal protein S27; RP11-422P24.3; rps27; rps27 {ECO:0000312; S27; S27-1; Small ribosomal subunit protein eS27 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 6232 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P42677 | |
| RPS27 | |
| Synthetic peptide sequence (PLAKDLLHPSPEE) corresponding to the N-terminus amino acids of RPS27 (aa 2-15). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction