missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ MPure-32 aNAP System
Shop All MP Biomedicals Products
Click to view available options
Quantity:
32 Samples
Unit Size:
Each
Beschreibung
- MPure-32™ aNAP System is a fully automated magnetic bead-based nucleic acid extraction system. It can process up to 32 samples simultaneously within a short period of time (around 40 to 60 minutes).
- Besides its capability to extract nucleic acid with high purity and yield, MPure-32™ aNAP System:
- Is easy to use
- Eliminates the risk of human error and cross-contamination
- Reduces working time
- Ensures accurate temperature control
- Is highly flexible
- MP Biomedicals also offers a range of reagent kits for various types of biological samples. The user has the flexibility to self-assemble the reagents with the relevant consumables for maximum cost-effectiveness or purchase the pre-assembled reagent kits for greater convenience.
- Specifications:
- Run Time: 45 to 60 minutes
- Samples per run: From 1 to 32 samples
- Weight (NW): 21 kg
- Dimensions: 38 x 35 x 37 cm
- Power Supply: 3.2 A, 100/240 V
- Processing Volume: 50 μL to 1,000 μL
- Magnetic Rod: > 4,300 gauss
- Temp Control: 1 set
- Heating Block: Yes (2 pcs)
- Heating: RT 70°C
- UV Lamp: Yes
- Display: 5.5 in. Touch Screen
Spezifikation
Spezifikation
| Sample Type | Tissue, Cell Culture, Bacteria |
| Suitable Downstream Analysis | PCR, Real-time PCR, Enzymatic Reactions |
| Volume (Metric) Processing | 50 to 1000 μL |
| Plates per Deck | 8 |
| Format | Tube |
| Quantity | 32 Samples |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur