Learn More
MP Biomedicals™ MPure™ Viral/Pathogen Nucleic Acids Extraction Kit B
For bacterial culture, swab samples and environmental samples
Brand: MP Biomedicals™ 117022130
Description
MPure Viral/Pathogen Nucleic Acids Extraction Kit B is used with the MPure-12 instrument for extraction of viral and bacterial DNA/RNA from bacterial culture, swab samples and environmental samples. Starting samples include clinical specimens such as sputum, bronchoalveolar lavage, cell-free body fluids, urine, respiratory specimens, bacterial culture either solid or in suspension and environmental samples such as soil, water and feces. The kit contains everything needed for sample preparation. However, for tough samples (soil or feces), sample preparation using FastPrep™ Instruments and Lysing Matrix Tubes can greatly increase yields as well as shorten processing times.
- Completely automated purification protocol
- Can be run in as little as 55 minutes
- Extracted nucleic acids can be used in downstream applications
Specifications
Store at room temperature | |
Bacteria, Virus, Swab | |
Nucleic Acid Purification; viral and bacterial DNA/RNA from cell-rich samples | |
48 Preps | |
As little as 55 minutes |
Viral/Pathogen Nucleic Acids Extraction Kit B | |
MPure-12 Automated Nucleic Acid Purification System | |
Proteinase K: 40μL; Lysis Buffer 2: 720μL; Binding Buffer 1: 720μL; Magnetic Bead Solution: 800μL; Washing Buffer 1: 1000μL; Washing Buffer 2: 1000μL; Washing Buffer 3: 1000μL; Elution Buffer 1: 1000μL; Elution Buffer 2: 1000μL; BL2 Buffer: 400μL | |
Bacteria Pellet: 100-200μL /Up to 109 bacteria (about OD600 = 3); Bacterial Colony: 100-200μL /1-3 colony; Swab samples: 100-200μL liquid transport media |