Learn More
Tocris Bioscience™ MRS 1220
Highly potent and selective hA3 antagonist
Brand:  Tocris Bioscience™ 1217/5
216.84 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
Product Code. 18757366
  • 275.00 € / 5mg
Explore more special offers
Description
Description
A potent and highly selective antagonist at the human A3 adenosine receptor (Ki values are 0.65, 305, and 52 nM at hA3, rA1 and rA2A respectively. Displays an IC50 value > 1 μM for inhibition of binding to rat A3 receptors).
Specifications
Specifications
5 mg
>98%
C21H14ClN5O2
N-[9-Chloro-2-(2-furanyl)[1,2,4]-triazolo[1,5-c]quinazolin-5-yl]benzene acetamide
Solid
Store at room temperature
183721-15-5
403.83
MRS 1220