missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MS 3.1 Standard Attachment

Product Code. 10473472 Shop All IKA Products
Click to view available options
No. of Positions:
1
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10473472

Brand: IKA™ 0003426300

Only null left
Add to basket
Edge
Add to basket
This item is not returnable. View return policy

Helps to hold vessels during the shaking process, suitable for a wide range of IKA™ shakers.

  • For test tubes and small vessels up to 50mm in diameter
  • Included with the minishakers MS 3 Basic and MS 3 digital
TRUSTED_SUSTAINABILITY

Specifications

Holds Test Tubes
Tube Size Up to 50 mm dia.
Product Type MS 3.1 Standard Attachment
For Use With (Equipment) Minishakers MS 3 Basic, MS 3 Digital
No. of Positions 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.