Learn More
Thermo Scientific™ MS(PEG)12 Methyl-PEG-NHS-Ester Reagent
Pegylate amines (-NH2 groups) on proteins (lysine residues) or assay surfaces using MS(PEG)n, amine-reactive (NHS ester), methyl-capped PEG compounds (4 to 24 EG units).
337.00 € - 978.00 €
Specifications
Chemical Reactivity | Amine |
---|---|
Content And Storage | Store desiccated at -20°C. |
Label Type | Polyethylene Glycol (PEG), Blocking Groups |
Reactive Moiety | Active Ester, Succinimidyl Ester, NHS Ester |
Spacer | 12 ethylene glycol units |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11801325
|
Thermo Scientific™
22685 |
100 mg |
337.00 €
100mg |
Estimated Shipment: 02-01-2025
Log in to see stock. |
|||||
10743295
|
Thermo Scientific™
22686 |
1 g |
978.00 €
1g |
Estimated Shipment: 06-01-2025
Log in to see stock. |
|||||
Description
Thermo Scientific™ Pierce MS(PEG)n reagents are methyl-terminated, polyethylene glycol compounds (n equals 4 to 24 PEG units) activated as NHS esters for covalent pegylation of primary amines on proteins (e.g., lysines) or assay surfaces.
MS(PEG)n is the abbreviation for a set of compounds having polyethylene glycol (PEG) spacers with methyl (—CH3) and amine-reactive NHS-ester groups at opposite ends. The unbranched, hydrophilic, discrete-length molecules have the form Methyl-PEGn-NHS Ester, where the subscript “n” denotes 4, 8, 12, or 24 ethylene glycol units. The N-hydroxysuccinimide (NHS) ester is spontaneously reactive with primary amines (—NH2), providing for efficient PEGylation of proteins, peptides and other amine-containing molecules or surfaces.Highlights:
- NHS-activated for efficient PEGylation of primary amines at pH 7-9; reaction of NHS-ester group results in formation of stable, irreversible amide bonds
- Fully characterized PEGylation reagents with defined PEG chain lengths; molecules of discrete molecular weight for consistency of performance in protein-modification applications
- Provided as a series of 4, 8, 12 and 24 ethylene glycol units, enabling modification procedures to be optimized for a specific application while retaining all the benefits associated with protein PEGylation
- PEG spacer provides unique advantages, including increased stability, reduced tendency toward aggregation and reduced immunogenicity
- Easy-to-follow instructions increase the likelihood of a successful outcome
Specifications
Amine | |
Polyethylene Glycol (PEG), Blocking Groups | |
12 ethylene glycol units | |
NHS-Ester Reagent |
Store desiccated at -20°C. | |
Active Ester, Succinimidyl Ester, NHS Ester | |
Water |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.