missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25028-1-AP-20UL
This item is not returnable.
View return policy
Description
MyoD-E or musculin-E heterodimers bind avidly to consensus E box motifs, which are functionally important elements in the promoter regions of many musclespecific terminal differentiation genes. MyoD complexes potently induce transcriptional activation, while musculin complexes bind adjacent to MyoD DNA-binding regions to represses MyoD activity, which then results in the delayed expression of muscle-specific genes. Musculin is highly expressed in undifferentiated and proliferating myoblasts in culture, and its expression is down regulated during myogenesis and at the onset of terminal differentiation.Specifications
MSC | |
Polyclonal | |
Unconjugated | |
MSC | |
ABF 1, ABF1, Activated B cell factor 1, bHLHa22, MSC, Musculin, MYOR | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17681, 9242 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O60682, O88940 | |
Msc | |
MSC Fusion Protein Ag13489 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |