missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MSH3 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22393-1-AP-150UL
This item is not returnable.
View return policy
Description
PUS1 converts uridine into pseudouridine after the nucleotide has been incorporated into RNA. Pseudouridine may have a functional role in tRNAs and may assist in the peptidyl transfer reaction of rRNAs.Specifications
MSH3 | |
Polyclonal | |
Unconjugated | |
MSH3 | |
Divergent upstream protein, DUC1, DUG, DUP, hMSH3, Mismatch repair protein 1, MRP1, MSH3, mutS homolog 3 (E. coli) | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
4437 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P20585 | |
MSH3 | |
MSH3 Fusion Protein Ag18060 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |