missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Velp Scientifica™ MSL 8 Digital High Volume Magnetic Stirrer
Digital magnetic stirrer with continuous operation at both low and high speeds with timer, auto-reverse and SpeedServo™. MSL 8 Digital High Volume Magnetic Stirrer is an ideal solution for large capacity laboratory stirring applications in life science and pharmaceuticals.
Brand: Velp Scientifica™ F203A0510
237.40 EUR valid until 2025-12-31
Use promo code "24299" to get your promotional price.
This item is not returnable.
View return policy
Description
- High-volume magnetic stirrer with digital display to accurately set working conditions
- Programmable operations with integrated timer and auto-reverse
- For high-volume preparations, cell and tissue cultures, and buffer preparations
- SpeedServo™ technology maintains a constant speed
- Unattended stirring from 1 min to 99 hrs and 59 min
- Brushless motor provides continuous operation for high volumes at any speed
- Robust stainless steel top for easy visualization of color changes and particles
Specifications
40 to 1500 rpm | |
Epoxy | |
Digital | |
110/240 V | |
Magnetic Stirrer |
1 | |
8 L | |
Square | |
5 W |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction