Learn More
Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent
Offer maximum volatility with the Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent.
Brand: Thermo Scientific™ TS-48915
183.14 EUR valid until 2025-03-14
Use promo code "24142" to get your promotional price.
Description
Use Thermo Scientific™ MSTFA and MSTFA + 1% TMCS Silylation Reagent for GC analyses of early eluting compounds that would otherwise be obscured in the chromatogram. MSTFA, the most volatile TMS-amide available, and its byproduct N-methyltrifluoroacetamide typically elute with the solvent front, allowing TMS derivatives of small molecules to be analyzed. Addition of Thermo Scientific™ TMCS aids derivatization of amides, secondary amines, and hindered hydroxyls not derivatized by MSTFA alone.
- Trimethylsilyl donor strength comparable to BSA and BSTFA
- Reacts to replace labile hydrogens on a wide range of polar compounds with a Si(CH3)3 group
- Used to prepare volatile and thermally stable derivatives for GC and MS
- Volatile byproduct, N-methyltrifluoroacetamide, has an even lower retention time than MSTFA
- Often TMS derivatives of small molecules can be analyzed when derivatized with MSTFA because the byproducts and the reagent itself usually elute with the solvent front
- Addition of Thermo Scientfic TMCS aids derivatization of amides, secondary amines and hindered hydroxyls not derivatized by MSTFA alone

Specifications
GC/MS | |
98 to 100, 0.1 to 1% | |
10 x 1 mL ampules |
Flammable, Irritant | |
2°C to 8°C | |
MSTFA plus 1% TMCS (2,2,2-Trifluoro-N-methyl-N-(trimethylsilyl)-acetamide, Chlorotrimethylsilane) |
Safety and Handling
- N-Methyl-N-(trimethylsilyl)trifluoroacetamide
Signal Word
- Warning
Hazard Category
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H226-Flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : Acetamide, 2, 2, 2-trifluoro-N-methyl-N-(trimethylsilyl)-
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.