missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MT 40.10 Grinding Chamber

Stainless steel beater reduces soft, medium, hard and brittle materials with a Mohs hardness of up to 5 (manganese or apatite: 5 Mohs).

Brand:  IKA™ 0004425000

110.35 EUR valid until 2025-04-30
Use promo code "24119" to get your promotional price.


Product Code. 15583824

  • 107.00 € / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chamber is made of transparent plastic (PP) so grinding tests can be observed at any time
  • 40mL useful volume
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Grinding Chamber
40 mL
Polypropylene
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
IKA™ MT 40.10 Grinding Chamber > Capacity: 40mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.