Learn More
MTAP Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11475-1-AP-150UL
This item is not returnable.
View return policy
Description
The MTAP antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MTAP. This antibody recognizes human, rat, mouse antigen. The MTAP antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
MTAP | |
Polyclonal | |
Unconjugated | |
150 μL | |
4507 | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence | |
367 μg/mL | |
Rabbit | |
Primary | |
Human, Rat, Mouse |