missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent.

189.27 € - 460.69 €

Specifications

For Use With GC
Hazards Irritant
Storage 2°C to 8°C
View More Specs

Products 2
Product Code Brand Standard Components Price Quantity & Availability  
Product Code Brand Standard Components Price Quantity & Availability  
11527680
View Documents
Thermo Scientific™
TS-48920
MTBSTFA (N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide)
189.27 €
Each
Please to purchase this item. Need a web account? Register with us today!
11597861
View Documents
Thermo Scientific™
TS-48927
MTBSTFA +1% TBDMS (N-Methyl-N-(tert-butyldimethylsilyl)trifluoroacetamide, Tertbutyldimetheylchlorosilane
460.69 €
Pack of 10
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Produce stable TBDMS derivatives for GC analysis with the Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent. MTBSTFA derivatizes hydroxyl, carboxyl, thiol and primary and secondary amines with typically >96% yields, and the reaction byproducts are volatile and neutral. TBDMS ethers are 104 times more stable to hydrolysis than the corresponding TMS ethers. Addition of Thermo Scientific™ TBDMCS increases silylation potential.

Additional Product Information:
  •  Derivatives have a high molecular concentration at M-57.
Specifications

Specifications

GC
Irritant
2°C to 8°C
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MTBSTFA and MTBSTFA + 1% TBDMCS Silylation Reagent

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.