missing translation for 'onlineSavingsMsg'
Learn More

IKA™ MTT 40.10 Disposable Grinding Chamber

Disposable grinding chamber equipped with a titanium beater.

Brand:  IKA™ 0020015380

Product Code. 16268501

  • 180.00 € / Pack of 10
Estimated Shipment: 20-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Volume: 40 mL
  • With titanium beater
Specifications

Specifications

Grinding Chamber
Tube Mill
0.395 kg
40 mL
10/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers