missing translation for 'onlineSavingsMsg'
Learn More

Mucic acid, 98%

Product Code. 10438480 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 10438480

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 526-99-8 | C6H10O8 | 210.138 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 526-99-8
Molecular Formula C6H10O8
Molecular Weight (g/mol) 210.138
MDL Number MFCD00004239
InChI Key DSLZVSRJTYRBFB-DUHBMQHGSA-N
Synonym galactaric acid, mucic acid, 2r,3s,4r,5s-2,3,4,5-tetrahydroxyhexanedioic acid, saccharolactic acid, mucicacid, unii-e149j5otif, d-galactaric acid, galactosaccharic acid, galactarate, meso-galactaric acid
PubChem CID 3037582
ChEBI CHEBI:30852
IUPAC Name (2S,3R,4S,5R)-2,3,4,5-tetrahydroxyhexanedioic acid
SMILES C(C(C(C(=O)O)O)O)(C(C(=O)O)O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color White to Beige
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Plastic bottle
Quantity 500 g
Beilstein 03, IV, 1292
Merck Index 15, 4361
Formula Weight 210.14
Percent Purity ≥97.5%
Physical Form Fine Crystalline Powder
Chemical Name or Material Mucic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.