missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Mueller Hinton Agar (Dehydrated)

Perform effortless Antimicrobial Susceptibility Testing (AST) while conforming to EUCAST and CLSI with this medium.

Brand:  Thermo Scientific™ CM0337R

Product Code. 12771500

  • 700.00 € / 2.50kg
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform effortless Antimicrobial Susceptibility Testing (AST) using Thermo Scientific™ Oxoid™ Mueller Hinton Agar (Dehydrated), a medium designed and manufactured according to the European standard, EUCAST1 and the American standard, CLSI2, 3, 4.

The major use of Mueller-Hinton Agar is for Antimicrobial Susceptibility Testing (AST). It has become the standard medium for the Kirby-Bauer method5, 6 and can be used in internationally recognized standard procedures.

  • Specified by the Clinical and Laboratory Standards Institute (CLSI) formerly the National Committee for Clinical Laboratory Standards (NCCLS) 2,3,4 and The European Committee on Antimicrobial Susceptibility Testing (EUCAST)1
  • Standard medium for the Kirby-Bauer method5, 6

The inclusion of starch in the medium ensures that toxic factors produced during microbial growth will be absorbed, and its presence is often essential to establish growth from very small inocula.

Antibiotic susceptibility tests are performed in accordance with and meet the acceptance limits of the current ISO/TS 16782 using the following microorganisms:

Staphylococcus aureus ATCC®25923 WDCM00034
Staphylococcus aureus ATCC®29213 WDCM00131
Staphylococcus aureus ATCC®43300 WDCM00211
Staphylococcus aureus NCTC 12493 WDCM00212
Escherichia coli ATCC®25922 WDCM00013
Escherichia coli ATCC®35218
Pseudomonas aeruginosa ATCC®27853 WDCM00025
Enterococcus faecalis ATCC®33186 WDCM00210
Enterococcus faecalis ATCC®29212 WDCM00087
Streptococcus pneumoniae ATCC®49619
Haemophilus influenzae ATCC®49247
Haemophilus influenzae ATCC®49766

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Conforms to CLSI standards. Industrial Reference: FDA.
Powder
Each
Mueller Hinton Agar
Agar
For 13.2 L mediμm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Laboratory Use Only