Learn More
Nabertherm™ Muffle Furnaces
Nabertherm furnaces manufactured from expertly finished, high quality materials and are easy to operate. The insulation out of non-classified fibre materials permits energy saving operation and fast heating times due to low heat storage and thermal conductivity.
4100.00 € - 7680.00 €
Product Code | Brand | Capacity (Metric) | Controller Type | Style | Temperature (Metric) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Controller Type | Style | Temperature (Metric) | Price | Quantity & Availability | ||
17347859
|
Nabertherm™
L-055K1ANL |
5 L | B510 | Flap Door | 1100°C |
4100.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17307879
|
Nabertherm™
L-095K1ANL |
9 L | B510 | Flap Door | 1100°C |
4330.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17387879
|
Nabertherm™
L-095K4ANL |
9 L | B510 | Flap Door | 1400°C |
6500.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17397879
|
Nabertherm™
L-095K4ANO |
9 L | C550 | Flap Door | 1100°C |
7290.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17307889
|
Nabertherm™
L-095H4ANL |
9 L | B510 | Lift Door | 1400°C |
6500.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17317889
|
Nabertherm™
L-095H4ANO |
9 L | C550 | Lift Door | 1400°C |
7290.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17327889
|
Nabertherm™
L-155H1ANL |
15 L | B510 | Lift Door | 1100°C |
5095.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17347899
|
Nabertherm™
L-155K4ANL |
15 L | B510 | Flap Door | 1400°C |
6890.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17357899
|
Nabertherm™
L-155K4ANO |
15 L | C550 | Flap Door | 1400°C |
7680.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17367899
|
Nabertherm™
L-155H4ANL |
15 L | B510 | Lift Door | 1400°C |
6890.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
17377899
|
Nabertherm™
L-155H4ANO |
15 L | C550 | Lift Door | 1400°C |
7680.00 €
Each |
Estimated Shipment: 27-01-2025
Log in to see stock. |
||
Description
- Heating from two sides by ceramic heating plates with integrated heating element which is safeguarded against fumes and splashing, and easy to replace
- Insulation made of non-classified fibre material
- Textured stainless steel sheet housing
- Equipped with either a flap door (L) or lift door (LT)
- Dual shell housing for low external temperatures and high stability
- Adjustable air inlet integrated into the door, exhaust air outlet in the rear wall of furnace
- Supplied with B510 controller with four programs and five freely programmable ramps, or C550 with 10 freely adjustable programs and 20 segments
- Both controllers have integrated USB interface and offer plain text display in different languages
- Alternative furnace sizes and temperature ranges are available on request
Specifications
Furnace | |
230 V |