missing translation for 'onlineSavingsMsg'
Learn More

Multi 7 ions, 10mg/L of each Component, for Ion Chromatography, H2O matrix, Fisher chemical™

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

149.00 € - 284.00 €

Specifications

Chemical Name or Material Multi 7 ions
Concentration 10mg/L of each Component
Packaging Bottle
Type Multi-component Standard Solution
Analyte or Component Names F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12932235
View Documents Promotion Details
Fisher Chemical
A 101 100
100 mL Bottle
149.00 €
100mL
Please to purchase this item. Need a web account? Register with us today!
12942235
View Documents Promotion Details
Fisher Chemical
A 101 500
500 mL Bottle
284.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ions (7 components): F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
Specifications

Specifications

Multi 7 ions
Bottle
F-,Cl-,NO2-,Br-,NO23-,PO43-,SO42
Liquid
10mg/L of each Component
Multi-component Standard Solution
H2O
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 24 Months. (12 Months after opening)

SDS
Documents

Documents

Product Certifications