Learn More
Thermo Scientific™ Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
Brand: Thermo Scientific™ 840-303600
This item is not returnable.
View return policy
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
4-position Carousel Base | |
4-position Carousel Base | |
GENESYS 140/150/180, BioMate 160 Spectrophotometers | |
Up to 4 10 mm Pathlength Cells |