Learn More
Thermo Scientific™ Multi-cell accessories for GENESYS™ 140/150/180 and BioMate™ 160 Spectrophotometers
Speed up your analysis with high-throughput measurements.
Brand:  Thermo Scientific™ 840-314700
 View more versions of this product
Product Code. 15871148
  • 431.00 € / Each
Explore more special offers
Description
Description
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurment requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Speed up your analysis with high-throughput measurements.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Accommodates four long-path rectangular cells up to 50mm. Please note: Not compatible with low volume cells (< 3.5mL) or any other type of cell holder.
4-position Carousel with Long Path Cell Holders
Up to 4 Long Pathlength Rectangular Cells (up to 50 mm)
4-position Carousel with Long Path Rectangular Cell Holders. Magnetic latch design for easy carousel exchange.
GENESYS 140/150/180, BioMate 160 Spectrophotometers