missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Multi-channel Cassette

For use with multi-channel pumps

Brand:  Heidolph™ 524-90021-00

249.85 EUR valid until 2024-11-30
Use promo code "21681" to get your promotional price.


 View more versions of this product

Product Code. 10239461

  • 277.00 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Flow rates vary by cassette size
  • Suitable for tubes with 0.9 or 1.6 mm wall thickness (wt)
Specifications

Specifications

0.24 to 27 mL/min.
Tubing with 1.6 mm wall thickness
Medium
Cassette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers