Learn More
Heidolph™ Multi-channel Pump Head
Increase your throughput by running more cassettes on one single pump drive and pump head system
Brand:  Heidolph™ 524-81220-00
 View more versions of this product
Product Code. 10504932
  • 4455.00 € / Each
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Separate metering into multiple vessels with different feed rates in a single operation by using different tubing dimensions to adjust flow rate
  • Save time - tubing change is done in just seconds
  • Change your cassettes easily, even during operation – there are no restrictions
  • Cassette adjustments and changes are simple, just click in place
  • No additional purchase of a pump drive needed: upgrade your single-channel pump drive with just one adaptor for multi-channel use and pick the head/cassette configuration that matches your application needs
Specifications
Specifications
12 small cassettes
8 rollers
C 12
Pump Head