missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Multi Function 3D Rotators
For continuous operation, timed operation or different combinations to ensure optimal mixing conditions for any application
815.00 €
Specifications
Orbit | 23 mm |
---|---|
Rotating Angle | 0 to 360° |
Load Bearing Capacity (Metric) | 1 kg |
Speed Range | 5 to 100 rpm |
Voltage | 100/240 V |
Includes: Dimpled rubber mat; 6-tube securing bands
Product Code | Brand | Plug Type | Certifications/Compliance | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Plug Type | Certifications/Compliance | Price | Quantity & Availability | ||||
15514080
|
Fisherbrand™
88861048 |
EU, UK, CN Plug | CE |
815.00 €
Each |
In Stock
Log in to see stock. |
||||
Description
- Rotation speed: 5 to 100 rpm for long wave motion
- Reciprocal rotation: 0°to 360° turning angle for long and medium way motion
- Vibration: 0° to 5° turning angle, programmable in bursts of 0 to 5 seconds, for short wave motion
- Fits neatly into small workspaces
- Operating temperature range of 5 to 40°C allows for use in refrigerators or non-CO2 incubators
- Continuous or timed operation with automatic switch-off
- Display will show last used settings, even after power has been turned off.
- Continue to run by last set speed and the remaining time when the power recovered.
- Audible and visual alarm: In timed mode, when the time reaches zero, alarm will sound 5 times, then the time window shows“End“ and flickers all the time. Alarm can be stopped by touch any key.
Warranty
2 years
Specifications
23 mm | |
1 kg | |
100/240 V | |
2.2 lb. | |
266 mm | |
|
0 to 360° | |
5 to 100 rpm | |
5°C to 40°C | |
306 mm | |
Immune precipitations, Affinity matrix , Trypsinisation, Gel straining and distaining, Antibody staining, Washes, Hybridizations, Southern blots, Western blots | |
2 Years |