missing translation for 'onlineSavingsMsg'
Learn More

Heidolph™ Multi Reax

Multi Reax is a versatile model with a 3 mm shaking orbit.

Brand:  Heidolph™ 545-10000-02

1428.36 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.


Product Code. 17343040

  • 2045.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • It can accommodate between 12 (16 to 32 mm diameter) and 26 (10 to 16 mm diameter) tubes
  • Reaction vessels with snap-on lids are also safely fixed
  • The 3 mm shaking orbit and a speed control between 150 and 2000 rpm guarantee excellent mixing results
  • The integrated timer (up to 999 minutes) enables the automatic termination of the shaking function
  • After the time has expired, a signal tone is emitted
Specifications

Specifications

230 V
UK Plug
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.