missing translation for 'onlineSavingsMsg'
Learn More

ZVG™ Multiclean™ Plus Cleaning Cloth Roll

This 3-ply cleaning cloth roll made from 100% recycled material supplies high absorbency, bonding, and water resistance. Dot-to-dot embossing allows for efficient cleaning.

Brand:  ZVG™ 11338-12

Product Code. 17863555

  • 76.10 € / 2 rolls
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Durable Multiclean™ cloths are the standard in many workshops, whether mounted on the wall or placed on a floor stand, they are always within reach for thorough surface cleaning. They are suitable for use in industry and crafts, such as machine maintenance.

In a novel manufacturing process, the individual layers of the cellulose cloth are bonded together at many points across the entire surface. This creates a strong and tear-resistant cloth. This process combines the advantages of a thick wiping cloth (stability) with those of a multi-layered cloth (high absorbency).

  • Ideal for high efficiency and safety, especially when wiping metal chips
  • Made of 100% recycled material
  • Highly absorbent
  • Bonded with dot-to-dot embossing
  • Resistant to moisture
  • Lint-free
  • High-volume
  • Each roll contains 500 tear-offs
Specifications

Specifications

Paper Towel
35 cm
37 cm
Blue
3-ply
13.8 in.
14.6 in.
Recycled Paper
Single-use
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.