missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories

Guarantee excellent dispensing results and day-to-day performance with reagent dispenser accessories.

Brand:  Thermo Scientific™ 24070297

478.98 EUR valid until 2024-12-31
Use promo code "22005" to get your promotional price.


 View more versions of this product

Product Code. 11560745

  • 652.00 € / Each
Estimated Shipment: 17-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Guarantee excellent dispensing results and day-to-day performance with Thermo Scientific™ Multidrop™ Combi, 384 and DW Reagent Dispenser Accessories.

Ordering Information:

  • Cat. No. 24070297 and 24070290 include tip combs
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Long Tubing Set
Long tubing set for standard dispensing cassette (tubing length 200 cm)
5 to 2500 μL
<24 mL
Autoclavable (50 times at 121°C, 1 bar, 20 min.)
Polypropylene Tip, Silicone Tubing
Multidrop Combi, Multidrop 384, Multidrop DW
Tip: 0.5 mm; Tubing: 1.3 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.