Learn More
Thermo Scientific™ Multidrop™ Combi Reagent Dispenser
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo ScientificTM MultidropTM Combi Reagent Dispenser.
20883.00 € - 34240.00 €
Specifications
Product Type Reagent Dispenser
Channels 8
View More Specs
Products 3
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
15377400
View Documents
Thermo Scientific™
5840320
Multidrop Combi with SMART 2 option This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
17801692
View Documents
Thermo Scientific™
5840330
Multidrop™ Combi+ Reagent Dispenser
30525.00 €
Each
In Stock
to see stock.
Add to basket
17811692
View Documents
Thermo Scientific™
5840340
Multidrop™ Combi SMART+ Reagent Dispenser
34240.00 €
Each
In Stock
to see stock.
Add to basket
Description
Description
Meet your lab's specific needs with outstanding flexibility and performance with the easy-to-use Thermo Scientific™ Multidrop™ Combi Reagent Dispenser, designed to support the varied requirements of pharmaceutical and biotechnology laboratories. It combines the most versatile features for reagent dispensing with excellent performance for drug discovery or genomic and proteomic assays.
The Multidrop Combi Reagent Dispenser handles a wide selection of plates and volume range and provides fast dispensing and high-throughput operation.
  • Provides precise dispensing over a 0.5 to 2500μL range, ensuring reproducible assay data
  • Accommodates microplates from 6 to 1536 wells and plate heights of 5 to 50mm
  • Visual icon-based graphic display makes it easy to use and program, even without training
  • Minimal dead volume and back-flushing reduces reagent costs
  • Easy to use Thermo Scientific FILLit Software provides increased flexibility and functionality
  • Full robotic compatibility gives increased throughput
  • Uses eight-channel detachable and autoclavable dispensing cassettes that are standard across the Multidrop range
Multidrop Combi SMART:
  • Adds an advanced built-in tracing system to the Multidrop Combi
  • Provides improved realiability and cassette lifetime traceability, greatly enhancing user efficiency and reporting capability
Recommended for:
  • Assay development
  • Primary and secondary screening
  • Compound storage
  • Genomics and proteomics research
  • PCR set-up
  • Sequencing set-up
  • Cell based assays
  • Bead based assays
  • ELISA assays
Specifications
Specifications
Reagent Dispenser
8