missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Multidrop™ DW Reagent Dispenser
Accelerate larger scale assays with the superior precision and accuracy of this high-speed reagent dispenser designed for repetitive dispensing of large volumes.
9785.00 € - 10420.00 €
Specifications
Product Type | Reagent Dispenser |
---|---|
Accuracy | 20µLl: ±2% (typical); 100µL: ±1% (typical); 900µL: ±1% (typical) |
Channels | 8 |
Volume (Metric) | 20 to 995 μL |
Increments | 5 μL |
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Voltage | Price | Quantity & Availability | |||||
10040894
|
Thermo Scientific™
5840170 |
200/240 V | N/A | ||||||
11550725
|
Thermo Scientific™
5840177 |
100/120 V | N/A | ||||||
Description
Accelerate larger scale assays with the superior precision and accuracy of the Thermo Scientific™ Multidrop™ DW Reagent Dispenser, a high-speed reagent dispenser designed for repetitive dispensing of large volumes in pharmaceutical and biotechnology laboratories.
Multidrop DW Reagent Dispenser portions various liquids into 96 standard and DW plates across 2a 0 to 995μL volume range.- Precise dispensing ensures high-quality assay results and reproducible data
- Wide volume range provides flexibility and increased speed
- Quick plate and volume selection, combined with easy maintenance, makes assay setup quick and easy
- Dispenser uses an eight-channel detachable and autoclavable dispensing cassette to ensure sterile conditions and to avoid cross contamination
- All reagent lines can be backflushed into the reagent bottle, minimizing the loss of expensive reagents
- For use with standard cassettes only
Includes:
One standard tube dispensing cassette (Cat. No. 24072670).
Recommended for:
- Cell based assays
- Compound storage
- Genomics and proteomics research
- ELISA assays
- Microbiological tests
Specifications
Reagent Dispenser | |
8 | |
5 μL | |
96-well plates; 96-deep well plates; 1.1mL tubes in 96 tube rack; Micronic 96-place tube racks | |
96-well plate: 20µL in 5 seconds; 50µL in 8 seconds; 300µL in 26 seconds; 900 µL in 74 seconds | |
12.2 in. | |
7.1 in. | |
12.6 in. | |
13.7 lb. | |
50/60 Hz |
20µLl: ±2% (typical); 100µL: ±1% (typical); 900µL: ±1% (typical) | |
20 to 995 μL | |
RS-232 | |
20μL: CV≤1.5% (typical), 100μL: CV≤1% (typical), 900μL: CV≤0.5% (typical) | |
<7 mL | |
31 cm | |
18 cm | |
32 cm | |
6.2 kg |