Learn More
WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter
Solve multiple measuring tasks at once and save time and costs. WTW™ MultiLine™ Multi 3620 IDS Multi-parameter Portable Meters feature two universal measuring channels for measuring pH, ORP, dissolved oxygen and/or turbidity simultaneously.
1705.00 € - 3360.00 €
Specifications
Channels | 2 |
---|---|
Display Type | LCD Color Graphic, Backlit |
IP Rating | IP 67 |
Memory | Manual 500 / 10000 automatic |
Power Supply | Wide range power supply, 4x 1.2 V NiMH battery or via Mini USB-B |
Product Code | Brand | Model | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | Price | Quantity & Availability | |||||
15568595
|
WTW™
2FD560 |
MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter |
1705.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15520476
|
WTW™
2FD56G |
MultiLine™ Multi 3620 IDS Multi-parameter Portable Meter, Set G |
3360.00 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Wireless ready
- Simultaneous display of all measured values
- Digital sensor recognition
- Two galvanically isolated measuring inputs
- Dual – 2 channels, 4 parameters
- Galvanic isolation of the sensor inputs prevents interferences of measuring signals
- Use the new wireless modules together with new IDS plug head sensors, be independent from cables and measure even at hardly accessible locations
- Efficient battery or mains supply, you will be able to measure cost-effectively in the field and in the lab
- Data transfer either takes place via USB stick or via an USB connection directly to a computer
- Color graphic display offers measured values as well as additional information such as sensor position, CMC and QSC
- Traceability of the individual measured values can be supported by activating the user administration
Scope of delivery includes the MultiLab™ Importer to import data via Excel™ as well as the MultiLab™ User (user administration).
Specifications
2 | |
IP 67 | |
Wide range power supply, 4x 1.2 V NiMH battery or via Mini USB-B |
LCD Color Graphic, Backlit | |
Manual 500 / 10000 automatic |