Learn More
Eppendorf™ Repeater™ M4 Starter Kit
Use this dispenser for precise and accurate dispensing of viscous solutions, contamination-free dispensing of toxic, radioactive and other hazardous liquids. Eppendorf™ Repeater™ M4 Starter Kit is ideal for repetitive dispensing of volumes in long series such as tube or plate filling.
Brand: Eppendorf™ 4982000314
390.15 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
Use this dispenser for precise and accurate dispensing of viscous solutions, contamination-free dispensing of toxic, radioactive and other hazardous liquids. Eppendorf™ Repeater™ M4 Starter Kit is ideal for repetitive dispensing of volumes in long series such as tube or plate filling.
- Includes Combitip Rack and Combitip Assortment Pack
- Dispenses volumes using the positive displacement principle. Liquid is directly dispensed without an air cushion, ensuring the correct volume regardless of the density, viscosity and volatility of the liquid
- Repetitive dispensing modes decrease the number of movements by allowing the user to fill the tip once and dispense up to 100 times without a refill
- Automatic Combitip recognition eliminates time consuming volume calculations
- Wide dispensing range: 1 to 10mL
- Stress-free work via integrated step counter: dispensing procedures can be continued error-free after an interruption or distraction
- Central Combitip ejector: fully emptied Combitip can be easily ejected with one hand
- Perfect for viscous or foaming solutions and liquids with high vapour pressure through positive displacement principle
- Safe handling of toxic, radioactive or infectious material
Specifications
CombiTip™ | |
Yes | |
1 to 10,000 μL | |
1 Each |
1 | |
Multipette M4 Starter Kit | |
Precise and accurate dispensing of viscous solutions, Contamination-free dispensing of toxic, radioactive and other hazardous liquids |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.