Learn More
Cytiva Multiphor II Buffer Strip Positioner, complete
Multiphor II Buffer Strip Positioner is a frame with slots that sits on top of ExcelGel precast gel after the gel is placed on Multiphor II cooling plate.
Brand:  Cytiva 80-6442-90
Product Code. 10279104
  • 559.00 € / Each
Estimated Shipment: 15-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The slots in the positioner facilitate placement of the buffer strips for electrophoresis and hold the strips securely in place during the run.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Only for use with Multiphor II cooling plate
Multiphor II Buffer Strip Positioner