Learn More
Gibco™ Murine Dendritic Cell Cytokine Package, PeproTech®
Brand:  Gibco™ MDC
608.03 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Product Code. 17823423
  • 912.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
Description
The Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of mouse dendritic cells.
IL-4 is a pleiotropic cytokine that regulates diverse T- and B-cell responses including cell proliferation, survival, and gene expression. Produced by mast cells, T cells, and bone marrow stromal cells, IL-4 regulates the differentiation of naive CD4+ T cells into helper Th2 cells.
GM-CSF is a hematopoietic growth factor that stimulates the development of neutrophils and macrophages and promotes the proliferation and development of early erythroid megakaryocytic and eosinophilic progenitor cells.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Functional Assay
Dendritic Cell Cytokine Package
Mouse
100 μg Murine IL-4 (Cat. No. 214-14-100 UG), 100 μg Murine IL-4 (Cat. No. 214-14-100 UG)• 100 μg Murine GM-CSF (Cat. No. 315-03-100 UG), 100 μg Murine GM-CSF (Cat. No. 315-03-100 UG)
1 Kit