Learn More
Epredia™ MX35 Premier™ Disposable Low-profile Microtome Blades
Section hard tissues and high-quality thin sections with high-performance with Epredia™ MX35 Premier™ disposable low-profile microtome blades.
Brand:  Epredia™ 3051835
 View more versions of this product
Product Code. 12074088
  • 172.00 € / Pack of 50
Explore more special offers
Description
Description
  • Sharper edge with increased durability for longer blade life
  • Premier Plus blades provide greater durability, superior sharpness for lower blade friction and longer blade life
Specifications
Specifications
Disposable Low Profile Microtome Blade
Designed for routine histology and are good for a variety of tissue types, including hard tissue
34°
0.24 mm
Low
Single-use
0.01 in.