Learn More
MP BIOMEDICALS Myco-Out Mycoplasma Removal Spray
Myco-Out Mycoplasma Removal Spray is easy to use, fast and non-toxic, highly effective, and features a broad antimicrobial spectrum that eliminates Mycoplasma and inactivates various microorganisms.
Specifications
Antibiotic Spectrum | Broad spectrum |
---|---|
Classification | Chemically-defined |
Form | Liquid |
Product Type | Mycoplasma Removal |
Reagent Type | Surfactant |
Description
Mycoplasma is a frequent contaminant of cell cultures. It is the smallest and the simplest prokaryotic organism without cell walls. Therefore, it can pass through 0.22 μm filters easily. Moreover, the common antibiotics which target the cell wall, such as penicillin or β-lactam antibiotics, are ineffective against mycoplasmas. Mycoplasma contamination can severely affect the state of cells, causing changes in gene expression and metabolic characteristics, decelerated cell growth, abnormal cell differentiation, and cell death. It also severely affects cellular functions.
MP Biomedicals Myco-Out Mycoplasma Removal Spray is a ready-to-use peptide surfactant, which can quickly, efficiently and non-toxically eliminate mycoplasma from working surroundings such as:
- Benches
- Biosafety cabinets
- Laminar flow cabinets
- Cell incubators
- Pipettes
- Liquid nitrogen tanks
- Door handles, etc.
It also has a broad-spectrum antimicrobial disinfection effect.
Features:
- Easy to Use: Myco-Out Mycoplasma Removal Spray simplifies Mycoplasma removal with a user-friendly, ready-to-use spray, eliminating the need for tedious methods like fumigation and alcohol wiping
- Fast and Non-Toxic: The mycoplasma-specific peptide surfactant derived from Bacillus subtilis works in just 5 to 10 minutes without harming cells
- Highly Effective: Efficacy studies confirm its effectiveness in removing Mycoplasma from surfaces like laminar flow cabinets coated with Mycoplasma-positive HeLa cell culture
- Broad Antimicrobial Spectrum: It eliminates Mycoplasma and inactivates various microorganisms, including bacteria, viruses, and fungal spores
- Safe to Use: The product is safe for users and the environment
Application Notes:
Myco-Out Mycoplasma Removal Spray is a ready-to-use peptide surfactant intended to:
- Quickly
- Efficiently
- and non-toxically eliminate mycoplasma from working surroundings
Usage Statement:
Research Use Only (RUO)
Specifications
Broad spectrum | |
Liquid | |
Surfactant |
Chemically-defined | |
Mycoplasma Removal | |
Non-sterile |
Safety and Handling
missing translation for 'shelfLife' : 18 Months
- Myco-Out Mycoplasma Removal Spray
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Isopropanol, ethanol