Learn More
Lonza™ Mycoalert™ Mycoplasma Assay Control Set
It is recommended to always include the Control Set to ensure that the operator and all of the assay reagents are performing properly. Mycoalert™ Mycoplasma Assay Control Set is designed for use with the MycoAlert™ Mycoplasma Detection Kit.
Brand:  Lonza™ LT07-518
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
Product Code. 11680271
  • 330.00 € / 10 tests
Explore more special offers
Description
Description
Convenient & fast
  • Simply add two reagents to the culture supernatant
  • Perform two luminescence readings
  • Results in < 20 min
  • Easy interpretation of results
Universal
  • Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
  • Negligible interference with media components
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Mycoalert assay control : 1 x 1mL, Mycoalert assay buffer: 1 x 2mL
2 to 8°C
Mycoplasma Detection
10 Tests
Mammalian Cells
Mycoplasma detection
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • MycoAlert Assay Control Set
Hazard Statement
  • EUH210-Safety data sheet available on request.