Learn More
Lonza™ Mycoalert™ Mycoplasma Detection Kit
Detect mycoplasma in culture. Mycoalert™ Mycoplasma Detection Kit is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.
855.00 € - 1830.00 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11650261
|
Lonza™
LT07-218 |
25 Tests |
855.00 €
25 tests |
Estimated Shipment: 16-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11630271
|
Lonza™
LT07-418 |
50 Tests |
1115.00 €
50 tests |
Estimated Shipment: 07-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10255393
|
Lonza™
LT07-318 |
100 Tests |
1830.00 €
1 set |
Estimated Shipment: 16-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Convenient & fast- Simply add two reagents to the culture supernatant
- Perform two luminescence readings
- Results in < 20 min
- Easy interpretation of results
- Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
- Negligible interference with media components
Specifications
Mammalian Cells | |
Mycoplasma Detection |