Learn More
Melford Mycophenolic Acid
Immunosuppressive agent.
Brand:  Melford M93000-0.1
Product Code. 30018547
  • 106.00 € / 100mg
Explore more special offers
Description
Description
Immunosuppressive agent.
Immunosuppressive agent. Suppresses cylokine induced nitric oxide production. Used to select animal cells expressing the E. Coli. gene for xanthine guanine phosphoribosyl transferase.
Specifications
Specifications
24280-93-1
C17H20O6
100 mg
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Melford Mycophenolic Acid > 100 mg