missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lonza™ MycoZap™ Mycoplasma Elimination Reagent
Antibiotic formulation to remove Mycoplasma from contaminated cell cultures
Specifications
Product Type | Mycoplasma Removal |
---|---|
For Use With (Application) | Mycoplasma elimination |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
11610281
|
Lonza™
LT07-818 |
1 treatment | 1 x 0.5 mL MycoZap™ Reagent 1, 1 x 1.5 mL MycoZap™ Reagent 2 |
N/A
|
N/A | ||||
11620281
|
Lonza™
LT07-918 |
5 treatments | 1 x 2.5 mL MycoZap™ Reagent 1, 1 x 7.5 mL MycoZap™ Reagent 2 |
N/A
|
N/A | ||||
Description
- Efficient mollicute elimination by a combination of antibiotic and antimetabolic agents.
- Minimal toxic effects.
- Eradicates Mycoplasma, Acholeplasma, Spiroplasma, and Entomoplasma.
- Suited for a broad range of cell cultures.
Specifications
Mycoplasma Removal | |
Mycoplasma elimination |