missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ UV Transilluminator Bulb, 365 nm, for myECL™ Imager
For use with Thermo Scientific myECL Imager
Brand: Thermo Scientific™ 62255
This item is not returnable.
View return policy
Description
Thermo Scientific™ myECL Imager Accessories and Replacement Parts are for use with Thermo Scientific myECL Imager.
Replacement parts for the myECL Imager and accessories to expand upon its capabilities.
Specifications
UV Transilluminator Bulb | |
myECL Imager | |
1 bulb |
UV Transilluminator Bulb, 365nm | |
myECL |
For Research Use Only. Not for use in diagnostic procedures.