missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myelin basic protein Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10458-1-AP-150UL
This item is not returnable.
View return policy
Description
10458-1-AP is capable of recognizing multiple isoforms of MBP.
The protein encoded by the classic MBP gene is a major constituent of the myelin sheath of oligodendrocytes and Schwann cells in the nervous system. However, MBP-related transcripts are also present in the bone marrow and the immune system. These mRNAs arise from the long MBP gene (otherwise called "Golli-MBP") that contains 3 additional exons located upstream of the classic MBP exons. Alternative splicing from the Golli and the MBP transcription start sites gives rise to 2 sets of MBP-related transcripts and gene products. The Golli mRNAs contain 3 exons unique to Golli-MBP, spliced in-frame to 1 or more MBP exons. They encode hybrid proteins that have N-terminal Golli aa sequence linked to MBP aa sequence. The second family of transcripts contain only MBP exons and produce the well characterized myelin basic proteins. This complex gene structure is conserved among species suggesting that the MBP transcription unit is an integral part of the Golli transcription unit and that this arrangement is important for the function and/or regulation of these genes.Specifications
Myelin basic protein | |
Polyclonal | |
Unconjugated | |
MBP | |
MBP, Myelin A1 protein, Myelin basic protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17196, 24547, 4155 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P02686, P02688, P04370 | |
Mbp | |
Myelin basic protein Fusion Protein Ag0713 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |