missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Myeloid Cell Marker Antibody (BM-1), Novus Biologicals™
Mouse Monoclonal Antibody
269.00 € - 472.00 €
Specifications
Antigen | Myeloid Cell Marker |
---|---|
Clone | BM-1 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL |
Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18439491
|
Novus Biologicals
NBP2-32880-0.1mg |
0.1 mg |
472.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18496791
|
Novus Biologicals
NBP2-32880-0.02mg |
0.02 mg |
269.00 €
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Myeloid Cell Marker Monoclonal antibody specifically detects Myeloid Cell Marker in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
Myeloid Cell Marker | |
0.2 mg/mL | |
Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
myeloid specific antigen, myeloid specific protein, p183 DNA-binding cytoplasmic ptotein | |
IgG1 κ | |
Protein A or G purified |
BM-1 | |
Flow Cytometry 1-2 ug/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
Human peripheral blood mononuclear cells | |
Primary | |
Store at 4C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title