Learn More
Thermo Scientific™ myImageAnalysis™ Software
myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
Brand: Thermo Scientific™ 62237
Description
Thermo Scientific™ myImageAnalysis Software is a full-featured, multifunction computer program to analyze and edit digital images of electrophoresis gels and blots that were acquired using gel documentation imagers or scanners.
The myImageAnalysis Software comprises a complete set of easy-to-use research tools for the quantitative digital analysis of image data from electrophoresis gels and Western blots. The software uses a powerful algorithm to automatically select and identify lanes and band-boundaries for calculation of migration distances or densitometry. Intuitive, ready-to-use functions (buttons) provide molecular weight determination, relative and absolute quantitation, and purity calculation. Images are easily loaded, viewed, compared, adjusted (e.g., contrast, rotation, crop) and annotated. The program reads and writes to standard file formats, providing compatibility with nearly any source image or final presentation method.Highlights:
Intuitive design sleek task-bars (ribbons) organize functions into four main tabs: File, Image, Lanes/Bands, Annotation
Compatible file types load and save images in popular file formats (TIFF, JPEG, PNG, etc.); analyze images from nearly any source, and easily share the results
Image comparison open two to six image files at once to review them simultaneously or toggle between them to edit them in parallel
Auto-identification accurate, customizable, automatic lane and band identification creates numbered, selectable objects for lane profile densitometry and analysis
MW determination versatile, trouble-free calculation of migration distances and molecular weights based on pre-installed or custom-added markers
Advanced quantitation automatically calculate sample purity based on band and lane intensities, or determine the relative or absolute quantities of bands, lanes or regions
Image refinement and annotation crop, straighten, invert and adjust (saturation, contrast, etc.); then add simple labels, notes and arrows
Exporting and sharing easily transfer data and images to Microsoft™ Excel™, Word™ and PowerPoint™ Programs for further analysis and presentation
Includes:
myImageAnalysis Software, 2 activation licenses, Quick Start Guide (within the software); user manual (within the software)
Requires:
Computer running MicroSoft™ Windows XP™, Windows Vista™, or Windows 7™ Operating System; not available for other operating systems at this time
Recommended for:
Automatically identify and delimit lane and band boundaries for analysis; Examine densitometry profiles of lanes and bands; Calculate and report migration distances and molecular weights of bands relative to standards; Overlay image of colored protein ladder onto image of chemiluminescent signal; Assess band purity by densitometry of stained bands relative to entire lanes; Measure and report the relative and absolute quantity of bands, lanes or regions; Quickly annotate and export analyzed images and quantitative data
Specifications
Software | |
myImageAnalysis | |
Contents: 1 software installation CD |
myECL Imager | |
2 licenses |