missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals myo-Inositol, Cell Culture Grade
Click to view available options
Quantity:
50 g
100 g
250 g
Description
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
Solubility
It is soluble in water (50 mg/ml).
Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
It has been tested with CHO, L929, and Vero cell lines and can be used for other cell culture applications. A method for HPLC identification and quantitation has been achieved by this chemical.
Solubility
It is soluble in water (50 mg/ml).
Notes
Solutions of myo-inositol can be autoclaved for use in cell culture. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under recommended storage conditions.
Specifications
Specifications
Melting Point | 222°C to 227°C |
Color | White |
Odor | Odorless |
Quantity | 100 g |
Beilstein | 1907329 |
Merck Index | 14,4978 |
Solubility Information | It is soluble in water (50mg/ml). |
Formula Weight | 180.16 |
Grade | Cell Culture |
Physical Form | Crystalline Powder |
Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction