missing translation for 'onlineSavingsMsg'
Learn More

MYO7A Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Invitrogen PA537182

Product Code. 15564281

  • 473.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This antibody detects endogenous protein at a molecular weight of 254 kDa. Purity is >95% by SDS-PAGE.

MYO7A Polyclonal antibody specifically detects MYO7A in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MYO7A
Polyclonal
Unconjugated
MYO7A
DFNA11; DFNB2; Hdb; motor protein; MyO VIIa; Myo7; Myo7a; myosin; myosin 7a; myosin VIIA; myosin VIIA (Usher syndrome 1B (autosomal recessive, severe)); myosin-VIIa; Myosin-VIIa motor domain; MYOVIIA; MYU7A; nmf371; NSRD2; polka; sh1; sh-1; shaker 1; unconventional myosin-VIIa; USH1B
Rabbit
Antigen affinity chromatography
RUO
4647
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Q13402
MYO7A
A synthetic peptide corresponding to residues in human Myosin VIIa
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.