Learn More
MYO7A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA1936
Description
PA1-936 detects myosin VIIa from mouse tissues as well as recombinant protein. PA1-936 has been successfully used in Western blot procedures. By Western blot, this antibody detects an ∽220 kDa protein representing myosin VIIa from mouse testes preparations. PA1-936 detects recombinant mouse myosin VIIa overexpressed in Sf9 insect cell lysate. PA1-936 immunizing peptide corresponds to amino acid residues 16-31 from mouse myosin VIIa. This sequence is completely conserved in mouse, porcine, and human myosin VIIa. PA1-936 immunizing peptide (Cat. # PEP-045) is available for use in neutralization and control experiments.
Myosin VIIa is a member of the myosin superfamily of actin-based motor proteins. Defects in the myosin VIIa gene are responsible for hearing impairment in shaker-1 (sh1) mice and causes Usher syndrome IB in humans. Usher syndrome associates congenital deafness, vestibular dysfunction, and retinitis pigmentosa and is the most common form of combined deafness and blindness. Structural features of myosin VIIa protein include an ATP binding N-terminal motor domain, a central region which possess five light-chain binding (IQ) motifs, and a C-terminal domain with three myosin tail homology (MyTH4) and talin-like homology regions.Specifications
MYO7A | |
Polyclonal | |
Unconjugated | |
MYO7A | |
DFNA11; DFNB2; Hdb; motor protein; MyO VIIa; Myo7; Myo7a; myosin; myosin 7a; myosin VIIA; myosin VIIA (Usher syndrome 1B (autosomal recessive, severe)); myosin-VIIa; Myosin-VIIa motor domain; MYOVIIA; MYU7A; nmf371; NSRD2; polka; sh1; sh-1; shaker 1; unconventional myosin-VIIa; USH1B | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
17921, 266714 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P97479 | |
MYO7A | |
Synthetic Peptide: S(16) G Q E F D V P I G A V V K L C(31) | |
200 μL | |
Primary | |
Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.